Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VOPP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038387.1 | XP_017893876.1 | 2 - 3 | 243 | 94 | |
GenBank mRNA | JO448084.2 | - | 2 - 3 | 242 | 94 | |
JO593067.2 | - | 2 - 3 | 253 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |