Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNTN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973695.2 | XP_013829149.1 | 2 - 3 | 192 | 73 | |
XM_013973696.2 | XP_013829150.1 | 2 - 3 | 192 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |