Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050011.1 | XP_017905500.1 | 4 - 5 | 1125 | 89 | |
GenBank mRNA | JT660139.1 | - | 1 - 2 | 484 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |