Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLAIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056479.1 | XP_017911968.1 | 1 - 2 | 134 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |