Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZMAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675290.3 | XP_005675347.1 | 2 - 3 | 502 | 99 | |
XM_018047588.1 | XP_017903077.1 | 2 - 3 | 579 | 99 | ||
XM_018047597.1 | XP_017903086.1 | 2 - 3 | 566 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |