Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676006.3 | XP_005676063.1 | 3 - 4 | 806 | 103 | |
GenBank mRNA | JT693932.1 | - | 5 - 6 | 1217 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |