Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056155.1 | XP_017911644.1 | 1 - 2 | 581 | 83 | |
XM_018056156.1 | XP_017911645.1 | 1 - 2 | 581 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |