Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005709457.3 | XP_005709514.2 | 2 - 3 | 224 | 65 | |
XM_013962893.2 | XP_013818347.1 | 2 - 3 | 221 | 65 | ||
GenBank mRNA | JO626692.2 | - | 2 - 3 | 205 | 65 | |
JT731603.1 | - | 2 - 3 | 168 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |