Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PALM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051196.1 | XP_017906685.1 | 6 - 7 | 571 | 63 | |
XM_018051197.1 | XP_017906686.1 | 6 - 7 | 455 | 63 | ||
XM_018051199.1 | XP_017906688.1 | 5 - 6 | 382 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |