Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APH1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677684.3 | XP_005677741.1 | 1 - 2 | 509 | 54 | |
GenBank mRNA | JO581146.1 | - | 1 - 2 | 439 | 54 | |
JT630049.1 | - | 242 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |