Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013963414.2 | XP_013818868.2 | 2 - 3 | 736 | 72 | |
GenBank mRNA | KA345547.1 | - | 1 - 2 | 163 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |