Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042768.1 | XP_017898257.1 | 6 - 7 | 1018 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |