Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677143.3 | XP_005677200.2 | 4 - 5 | 570 | 71 | |
GenBank mRNA | JO612023.2 | - | 3 - 4 | 1569 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |