Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C4BPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005690425.3 | XP_005690482.1 | 5 - 6 | 948 | 115 | |
XM_013970049.2 | XP_013825503.1 | 6 - 7 | 1044 | 115 | ||
XM_018060113.1 | XP_017915602.1 | 5 - 6 | 848 | 115 | ||
GenBank mRNA | JO421720.2 | - | 5 - 6 | 772 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |