Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018041137.1 | XP_017896626.1 | 1 - 2 | 241 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |