Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699651.2 | XP_005699708.1 | 3 - 4 | 427 | 59 | |
XM_005699652.2 | XP_005699709.1 | 2 - 3 | 304 | 59 | ||
XM_005699653.2 | XP_005699710.1 | 3 - 4 | 408 | 59 | ||
XM_018042779.1 | XP_017898268.1 | 2 - 3 | 279 | 59 | ||
GenBank mRNA | JO334438.2 | - | 3 - 4 | 215 | 59 | |
JO334442.2 | - | 3 - 4 | 215 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |