Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOGAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676609.3 | XP_005676666.1 | 4 - 5 | 818 | 120 | |
GenBank mRNA | JT672012.1 | - | 1 - 2 | 180 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |