Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679333.3 | XP_005679390.1 | 5 - 6 | 516 | 76 | |
GenBank mRNA | JO376554.2 | - | 6 - 7 | 426 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |