Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683655.3 | XP_005683712.1 | 7 - 8 | 835 | 91 | |
XM_018051947.1 | XP_017907436.1 | 8 - 9 | 1075 | 91 | ||
XM_018051948.1 | XP_017907437.1 | 9 - 10 | 1224 | 91 | ||
XM_018051949.1 | XP_017907438.1 | 7 - 8 | 943 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |