Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXNL4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692203.3 | XP_005692260.1 | 2 - 3 | 406 | 82 | |
GenBank mRNA | JO601951.2 | - | 2 - 3 | 939 | 82 | |
KA308423.1 | - | 2 - 3 | 341 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |