Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAPC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050991.1 | XP_017906480.1 | 4 - 5 | 764 | 66 | |
XM_018050992.1 | XP_017906481.1 | 4 - 5 | 936 | 66 | ||
GenBank mRNA | JO629970.2 | - | 4 - 5 | 758 | 66 | |
JO629971.2 | - | 4 - 5 | 758 | 66 | ||
KA307346.1 | - | 4 - 5 | 868 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Capra hircus Probe spans exons |