Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052670.1 | XP_017908159.1 | 7 - 8 | 859 | 62 | |
XM_018052671.1 | XP_017908160.1 | 7 - 8 | 859 | 62 | ||
XM_018052672.1 | XP_017908161.1 | 6 - 7 | 772 | 62 | ||
XM_018052673.1 | XP_017908162.1 | 7 - 8 | 859 | 62 | ||
XM_018052674.1 | XP_017908163.1 | 7 - 8 | 859 | 62 | ||
GenBank mRNA | KA314190.1 | - | 7 - 8 | 824 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |