Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005676357.3 | XP_005676414.1 | 2 - 3 | 249 | 95 | |
XM_018060985.1 | XP_017916474.1 | 2 - 3 | 164 | 95 | ||
GenBank mRNA | JO529858.2 | - | 2 - 3 | 255 | 95 | |
JO610473.2 | - | 1 - 2 | 288 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Capra hircus Probe spans exons |