Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039002.1 | XP_017894491.1 | 2 - 3 | 956 | 86 | |
XM_018039003.1 | XP_017894492.1 | 2 - 3 | 1216 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Capra hircus Probe spans exons |