Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HECA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053273.1 | XP_017908762.1 | 2 - 3 | 1459 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |