Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST6GAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055319.1 | XP_017910808.1 | 5 - 6 | 1666 | 58 | |
GenBank mRNA | JO637296.2 | - | 4 - 5 | 1304 | 58 | |
JO637297.2 | - | 4 - 5 | 1304 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |