Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLBA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066466.1 | XP_017921955.1 | 2 - 3 | 716 | 102 | |
XM_018066467.1 | XP_017921956.1 | 2 - 3 | 1442 | 102 | ||
XM_018066468.1 | XP_017921957.1 | 1 - 2 | 1134 | 102 | ||
GenBank mRNA | JO619024.2 | - | 1 - 2 | 352 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |