Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SOX30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051717.1 | XP_017907206.1 | 6 - 7 | 1930 | 98 | |
XM_018051718.1 | XP_017907207.1 | 4 - 5 | 2223 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |