Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692127.3 | XP_005692184.2 | 4 - 5 | 743 | 73 | |
XM_018062109.1 | XP_017917598.1 | 5 - 6 | 1743 | 73 | ||
XM_018062110.1 | XP_017917599.1 | 4 - 5 | 1196 | 73 | ||
GenBank mRNA | JO576941.1 | - | 1 - 2 | 886 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |