Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOMER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972981.2 | XP_013828435.1 | 8 - 9 | 1162 | 71 | |
XM_018066348.1 | XP_017921837.1 | 8 - 9 | 1036 | 71 | ||
GenBank mRNA | JO392641.2 | - | 1 - 2 | 1010 | 71 | |
KA336551.1 | - | 1 - 2 | 1213 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Capra hircus Probe spans exons |