Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674900.2 | XP_005674957.2 | 5 - 6 | 715 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |