Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO330223.2 | - | 3 - 4 | 2257 | 89 | |
JO330224.2 | - | 3 - 4 | 2257 | 89 | ||
JO368717.2 | - | 1 - 2 | 173 | 89 | ||
JO368718.2 | - | 1 - 2 | 173 | 89 | ||
JO427316.1 | - | 3 - 4 | 2304 | 89 | ||
JO427541.1 | - | 3 - 4 | 2304 | 89 | ||
JO427617.1 | - | 3 - 4 | 2304 | 89 | ||
KA341110.1 | - | 2 - 3 | 419 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |