Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HGH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058713.1 | XP_017914202.1 | 4 - 5 | 1609 | 77 | |
GenBank mRNA | JO428056.1 | - | 4 - 5 | 1039 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |