Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678537.3 | XP_005678594.2 | 7 - 8 | 1090 | 76 | |
XM_005678538.3 | XP_005678595.2 | 7 - 8 | 1058 | 76 | ||
XM_018043596.1 | XP_017899085.1 | 7 - 8 | 1090 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |