Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRNIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051608.1 | XP_017907097.1 | 6 - 7 | 581 | 66 | |
XM_018051609.1 | XP_017907098.1 | 6 - 7 | 576 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |