Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WFDC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018056874.1 | XP_017912363.1 | 3 - 4 | 330 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |