Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LY6G6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696575.3 | XP_005696632.1 | 2 - 3 | 250 | 76 | |
GenBank mRNA | KA327810.1 | - | 1 - 2 | 660 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |