Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678638.3 | XP_005678695.2 | 3 - 4 | 1159 | 65 | |
XM_018042342.1 | XP_017897831.1 | 4 - 5 | 842 | 65 | ||
XM_018042352.1 | XP_017897841.1 | 4 - 5 | 1236 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |