Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCLAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043911.1 | XP_017899400.1 | 3 - 4 | 387 | 91 | |
XM_018043912.1 | XP_017899401.1 | 2 - 3 | 110 | 91 | ||
XM_018043913.1 | XP_017899402.1 | 2 - 3 | 234 | 91 | ||
XM_018043914.1 | XP_017899403.1 | 3 - 4 | 387 | 91 | ||
XM_018043915.1 | XP_017899404.1 | 3 - 4 | 387 | 91 | ||
XM_018043916.1 | XP_017899405.1 | 3 - 4 | 387 | 91 | ||
GenBank mRNA | JT651496.1 | - | 3 - 4 | 256 | 91 | |
JT705388.1 | - | 2 - 3 | 228 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |