Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064581.1 | XP_017920070.1 | 2 - 3 | 227 | 78 | |
XM_018064582.1 | XP_017920071.1 | 2 - 3 | 217 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Capra hircus Probe spans exons |