Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102175358 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040763.1 | XP_017896252.1 | 1 | 1158 | 120 | |
GenBank mRNA | JO333133.2 | - | 1 | 1096 | 120 |
Gene Symbol: HMGN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051535.1 | XP_017907024.1 | 6 | 1363 | 120 | |
GenBank mRNA | KA321215.1 | - | 1 | 71 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Capra hircus Both primers and probe map within a single exon |