Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NR2C2AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018051482.1 | XP_017906971.1 | 5 | 1137 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon |