Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plpp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173003.1 | NP_001166474.1 | 5 - 6 | 728 | 102 | |
XM_005003770.3 | XP_005003827.2 | 5 - 6 | 1129 | 102 | ||
XM_013152814.2 | XP_013008268.1 | 4 - 5 | 980 | 102 | ||
GenBank mRNA | AF088283.1 | - | 5 - 6 | 725 | 102 | |
AF088284.1 | - | 5 - 6 | 728 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Cavia porcellus Probe spans exons |