Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172880.1 | NP_001166351.1 | 6 - 7 | 922 | 65 | |
NM_001172881.1 | NP_001166352.1 | 6 - 7 | 910 | 65 | ||
NM_001172882.1 | NP_001166353.1 | 6 - 7 | 953 | 65 | ||
NM_001172883.1 | NP_001166354.1 | 6 - 7 | 916 | 65 | ||
NM_001172884.1 | NP_001166355.1 | 6 - 7 | 915 | 65 | ||
NM_001172885.1 | NP_001166356.1 | 6 - 7 | 851 | 65 | ||
XM_005005812.3 | XP_005005869.2 | 6 - 7 | 949 | 65 | ||
XM_005005814.3 | XP_005005871.1 | 6 - 7 | 949 | 65 | ||
XM_005005816.3 | XP_005005873.1 | 6 - 7 | 949 | 65 | ||
XM_013157817.2 | XP_013013271.1 | 6 - 7 | 948 | 65 | ||
GenBank mRNA | D49416.1 | - | 6 - 7 | 916 | 65 | |
D49417.1 | - | 6 - 7 | 910 | 65 | ||
D49418.1 | - | 6 - 7 | 922 | 65 | ||
D49419.1 | - | 6 - 7 | 953 | 65 | ||
D49421.1 | - | 6 - 7 | 915 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Cavia porcellus Probe spans exons |