Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npy5r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001173414.1 | NP_001166885.1 | 1 | 1151 | 101 | |
XM_005008001.3 | XP_005008058.1 | 4 | 1367 | 101 | ||
XM_005008002.3 | XP_005008059.1 | 2 | 1575 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Cavia porcellus Both primers and probe map within a single exon |