Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ambp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172836.1 | NP_001166307.1 | 3 - 4 | 354 | 86 | |
XM_013142665.2 | XP_012998119.1 | 3 - 4 | 636 | 86 | ||
GenBank mRNA | AB006444.1 | - | 3 - 4 | 354 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Cavia porcellus Probe spans exons |