Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZnT35C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_135897.5 | NP_609741.3 | 7 - 8 | 1389 | 66 | |
NM_165105.3 | NP_723901.1 | 6 - 7 | 1461 | 66 | ||
GenBank mRNA | AY071460.1 | - | 7 - 8 | 1386 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |