Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275298.1 | NP_001262227.1 | 2 - 3 | 531 | 56 | |
NM_141170.3 | NP_649427.1 | 3 - 4 | 631 | 56 | ||
GenBank mRNA | AY047574.1 | - | 2 - 3 | 485 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |