Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nompA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299383.1 | NP_001286312.1 | 7 - 8 | 1412 | 57 | |
NM_080092.4 | NP_524831.2 | 7 - 8 | 1412 | 57 | ||
NM_165821.4 | NP_725030.2 | 7 - 8 | 1412 | 57 | ||
NM_165822.4 | NP_725031.2 | 6 - 7 | 1283 | 57 | ||
NM_206096.3 | NP_995818.1 | 6 - 7 | 1283 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |