Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299424.1 | NP_001286353.1 | 9 - 10 | 5000 | 60 | |
NM_078992.3 | NP_523716.2 | 9 - 10 | 5027 | 60 | ||
NM_165913.2 | NP_725185.1 | 9 - 10 | 4574 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |